Learn More
TARGETMOL CHEMICALS INC Eribaxaban 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3166810MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Eribaxaban (PD348292) is an orally available factor Xa inhibitor for the prevention and treatment of venous thromboembolism. Purity 99.74%
Catalog No. 50-287-6314