Learn More
TARGETMOL CHEMICALS INC Eribulin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T136881MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.Eribulin (B1939) is a nonpaclitaxel microtubule kinetic inhibitor with anticancer activity that inhibits polymerization of microtubule protein subunits by preventing lengthening and shortening of microtubules during cell division.Eribulin is used in the study of metastatic breast cancer and solid tumors. Purity 99.96%
Catalog No. 50-287-6930