Learn More
Medchemexpress LLC Eribulin-d3 mesylate | 00-00-0 | MFCD22572760 | 97.0% | 829.02 g·mol⁻¹ | C41H60D3NO14S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13442AS5MG
Encompass_Preferred
Eribulin-d3 (mesylate) is a deuterium-labeled form of eribulin mesylate supplied as the methanesulfonate salt. It is used as an isotopically labeled reference standard in analytical and preclinical research to study microtubule-targeting anticancer activity and to support quantitative mass spectrometry workflows.
  • deuterium-labeled internal standard for mass spectrometry
  • suitable for pharmacokinetic and metabolic tracer studies
  • supplied as the mesylate (methanesulfonate) salt
  • reported purity 97.0%
  • molecular weight approximately 829.02 g·mol⁻¹
Catalog No. 50-002-22054
Add to Cart
Request a Quote