Learn More
Selleck Chemical LLC Eribulin Mesylate E7389-1mg
SDP
Supplier:  Selleck Chemical LLC S89121MG
Encompass_Preferred
Eribulin Mesylate, an analogue of the marine natural product halichondrin B, is a nontaxane completely synthetic microtubule inhibitor.
Catalog No. 50-238-9824