Learn More
Apexbio Technology LLC Eriocitrin 13463-28-0 20mg
SDP
Supplier:  Apexbio Technology LLC N156320
Encompass_Preferred
Eriocitrin (CAS 13463-28-0) is a flavone glycoside predominantly isolated from lemons Experimental studies indicate that eriocitrin administration prior to acute exercise supports the maintenance of hepatic glutathione redox balance In its absence a pronounced shift toward glutathione oxidation is observed suggesting a role in cellular antioxidant defense This compound is thus of interest for research on oxidative stress regulation and may aid in elucidating protective mechanisms against exercise-induced oxidative imbalance
Catalog No. 50-000-90654