Learn More
Medchemexpress LLC Eriocitrin | 13463-28-0 | 596.53 | 1 MG

Supplier: Medchemexpress LLC HYN06361MG
Eriocitrin is a flavonoid isolated from lemons that functions as a powerful antioxidant. It inhibits the proliferation of liver cancer cells by arresting the cell cycle in the S phase and induces apoptosis by activating intrinsic signaling pathways involving mitochondria. Additionally, Eriocitrin attenuates acute kidney injury, reducing renal tissue cell apoptosis, inflammatory response, and oxidative stress.
- Isolated from lemons
- Powerful antioxidant properties
- Inhibits liver cancer cell proliferation
- Induces apoptosis
- Attenuates acute kidney injury in vivo
- Purity of 99.84%
- Appears as a solid, light yellow to yellow color
- Soluble in DMSO and Methanol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.