Learn More
Medchemexpress LLC Eriocitrin | 13463-28-0 | 596.53 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN063610MM/1ML
Encompass_Preferred
Eriocitrin is a flavonoid isolated from lemons that is a powerful antioxidant. It inhibits the proliferation of liver cancer cells by arresting the cell cycle in the S phase through upregulating p53, cyclin A, cyclin D3, and CDK6. This compound also triggers apoptosis by activating intrinsic signaling pathways involving mitochondria.
  • Powerful antioxidant
  • Inhibits liver cancer cell proliferation
  • Arrests cell cycle in S phase
  • Upregulates p53, cyclin A, cyclin D3, and CDK6
  • Triggers apoptosis via intrinsic signaling pathways
Catalog No. 50-003-88587