Learn More
Medchemexpress LLC Eriocitrin | 13463-28-0 | 596.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0636100MG
Encompass_Preferred
Eriocitrin is a flavonoid isolated from lemons that acts as a powerful antioxidant. It inhibits the proliferation of liver cancer cells by arresting the cell cycle in the S phase through the upregulation of p53, cyclin A, cyclin D3, and CDK6. Additionally, Eriocitrin induces apoptosis by activating intrinsic signaling pathways involving mitochondria.
  • Acts as a powerful antioxidant.
  • Inhibits the proliferation of liver cancer cells.
  • Induces apoptosis by activating intrinsic signaling pathways.
  • Upregulates p53, cyclin A, cyclin D3, and CDK6 levels in HepG2 cells.
  • Attenuates acute kidney injury induced by ischemia reperfusion in rats.
Catalog No. 50-003-88595