Learn More
Medchemexpress LLC Eriocitrin | 13463-28-0 | 596.53 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0636R5MG
Encompass_Preferred
Eriocitrin (Standard) is the analytical standard of Eriocitrin, a flavonoid isolated from lemons known for its powerful antioxidant properties. It is intended for research and analytical applications. This compound inhibits the proliferation of liver cancer cells by arresting the cell cycle in the S phase through the upregulation of p53, cyclin A, cyclin D3, and CDK6. Additionally, it triggers apoptosis by activating intrinsic signaling pathways involving mitochondria.
  • Analytical standard grade, serving as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Inhibits proliferation of liver cancer cells
  • Triggers apoptosis by activating intrinsic signaling pathways
Catalog No. 50-003-89094