Learn More
Medchemexpress LLC Eriocitrin | 13463-28-0 | 596.53 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN063650MG
Encompass_Preferred
Eriocitrin is a flavonoid isolated from lemons and functions as a powerful antioxidant. It inhibits the proliferation of liver cancer cells by arresting the cell cycle in the S phase, achieved by upregulating p53, cyclin A, cyclin D3, and CDK6. Additionally, Eriocitrin induces apoptosis by activating intrinsic signaling pathways involving mitochondria.
  • Isolated from lemons.
  • Functions as a powerful antioxidant.
  • Inhibits proliferation of liver cancer cells.
  • Induces apoptosis through intrinsic signaling pathways.
  • Attenuates acute kidney injury in rats.
  • High purity of 99.84%.
  • Appears as a solid, light yellow to yellow in color.
Catalog No. 50-003-88621