Learn More
TARGETMOL CHEMICALS INC Eriodictyol-7-O-glucoside 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN16211MG
Encompass_Preferred
Also available in 5 mg and bulk. Please contact Fisher for quotes. Eriodictyol-7-O-glucoside is an Nrf2 activator conferring protection against Cisplatin-induced toxicity. Eriodictyol-7-O-glucoside is a flavonoid and a free radical scavenger with antioxidant activity in pistachio skin. Purity 99.53%
Catalog No. 50-287-7047