Learn More
Santa Cruz Biotechnology ERK 2 Antibody (6H3) 50 µg/0.5 ml
Supplier:  Santa Cruz Biotechnology SC81459
Encompass
ERK 2 Antibody (6H3) is a mouse monoclonal IgG1, cited in 10 publications, provided at 50 µg/0.5 mlraised against the N-terminus of ERK 2 of human originrecommended for detection of ERK 2 of mouse, rat, human and canine origin by WB and IPSee ERK 2 (D-2): sc-1647 for ERK 2 antibody conjugates, including AC, HRP, FITC, PE, Alexa Fluor® 488, 594, 647, 680 and 790.At present, we have not yet completed the identification of the preferred secondary detection reagent(s) for ERK 2 Antibody (6H3). This work is in progress.
Catalog No. NC3957552