Learn More
Cayman Chemical ERK InhIbItr 500ug
SDP
Supplier:  Cayman Chemical 15944500 UG
Encompass_Preferred
A cell-permeable inhibitor that binds ERK2 near its docking domain (KD = 5 µM); blocks ERK-specific phosphorylation of Rsk-1 and TCF/Elk-1 but does not affect ERK1/2 phosphorylation by MEK1/2; completely prevents proliferation of several cancer cell lines
Catalog No. 50-297-0768