Learn More
Medchemexpress LLC Erk5-in-1 | 1234479-76-5 | >98.0% | 459.54 g/mol | C25H29N7O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY144035MG
Encompass_Preferred
ERK5-IN-1 is a small-molecule kinase inhibitor selective for ERK5 (MAPK7) with reported biochemical potency (ERK5 IC50 = 87 ± 7 nM) and activity against LRRK2[G2019S] (IC50 = 26 nM). It is intended for research use in biochemical and cell signaling studies of the MAPK/ERK pathway.
  • Potent ERK5 inhibition (IC50 ≈ 87 nM).
  • Also inhibits LRRK2[G2019S] (IC50 ≈ 26 nM).
  • Suitable for biochemical kinase assays and cell signaling studies.
  • Available as solid powder and as DMSO solution formats for convenience.
  • Molecular formula C25H29N7O2; molecular weight 459.54 g/mol.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-22772