Learn More
Medchemexpress LLC ERK5-IN-2 | 1888305-96-1 | 99.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12834150MG
Encompass_Preferred
ERK5-IN-2 is an orally active, selective ERK5 inhibitor (IC50s of 0.82 μM for ERK5 and 3 μM for ERK5 MEF2D) that does not interact with the BRD4 bromodomain. It has been shown to suppress tumor xenograft growth and bFGF-driven angiogenesis. This compound exhibits favorable pharmacokinetic properties including low intrinsic clearance and high oral bioavailability.
  • Selective ERK5 inhibitor with sub-micromolar IC50s
  • Does not interact with the BRD4 bromodomain
  • Suppresses tumor xenograft growth
  • Suppresses basic fibroblast growth factor (bFGF) driven angiogenesis
  • Exhibits low intrinsic clearance and high flux
  • Demonstrates low efflux ratio in caco-2 cell permeability assays
  • Significantly reduces tumor volumes in animal models
  • Features an oral bioavailability of 68%
Catalog No. 50-003-39528