Learn More
Chemglass Life Sciences Flask, 1000mL, Erlenmeyer, Heavy Duty Rim, Neck Fits # 11 Rubber Stopper
SDP
Supplier:  Chemglass Life Sciences CG85101L
Encompass_Preferred
Flask, 1000mL, Erlenmeyer, Heavy Duty, Neck Fits # 11 Rubber Stopper, Pack of 6
Catalog No. NC0484174