Learn More
Alkali Scientific Erlenmeyer Shake Flask, PC Conical, Flat Bottom, Sterile, 125mL, 24/Case, Vented Cap
SDP
Supplier:  Alkali Scientific EFN801
Encompass
• PC Bottle• Vent caps with 0.2 μm hydrophobic filters for gas exchange without contamination• Individually packaged in sterile bags|• Sterilized by E-beam, SAL=10^-6• Non-Pyrogenic, DNase/Rnase free
Catalog No. 50-302-7590