Learn More
Selleck Chemical LLC Erlotinib HCl S1023-1g
SDP
Supplier:  Selleck Chemical LLC S10231G
Encompass_Preferred
Erlotinib HCl is an EGFR inhibitor with IC50 of 2 nM in cell-free assays 1000-fold more sensitive for EGFR than human c-Src or v-Abl
Catalog No. 50-311-1251