Learn More
Medchemexpress LLC Erlotinib (hydrochloride) | 183319-69-9 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1200850MG
Encompass_Preferred
Erlotinib hydrochloride is a small-molecule epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor supplied as the hydrochloride salt for research use in biochemical and cell-based assays.
  • Inhibits EGFR tyrosine kinase with low-nanomolar potency.
  • Supplied as a solid hydrochloride salt suitable for analytical and biological studies.
  • High purity (99.9% by LCMS) for reliable experimental results.
  • Soluble in DMSO for preparation of stock solutions.
  • Commonly used in kinase assays and cellular signaling research.
Catalog No. 50-002-33959