Learn More
Cayman Chemical Erlotinib Inhibitors
SDP
Supplier:  Cayman Chemical 104835 G
Encompass_Preferred
An EGFR inhibitor (Ki = 2.7 nM); selective for EGFR over Src and Abl at 0.1 µM; induces caspase-3 and -7 activity in NCI H358 cells at 1 µM; reduces EGFR autophosphorylation in an HN-5 mouse xenograft model at 25 mg/kg
Catalog No. 50-288-9544