Learn More
Medchemexpress LLC ErSO-DFP | 2768139-76-8 | 99.6% | C20H17F5N2O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14407010MG
Encompass_Preferred
ErSO-DFP is a small-molecule anticipatory unfolded protein response (a-UPR) activator with enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells. It demonstrates antitumor activity in animal models and can cross the blood-brain barrier, supporting its use in both in vitro and in vivo research into ERα+ cancer biology and UPR mechanisms.
  • Activates the anticipatory unfolded protein response (a-UPR).
  • Shows enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells.
  • Demonstrates antitumor activity in mouse models.
  • Capable of crossing the blood-brain barrier.
  • Reported high purity (99.6%), suitable for research applications.
  • Available in multiple small pack sizes for laboratory use.
Catalog No. 50-002-58592