Learn More
Medchemexpress LLC ErSO-TFPy | 3035547-78-2 | C19H13F7N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1534685MG
Encompass_Preferred
ErSO-TFPy (TEQ103) is an ERα+ tumor cell inhibitor that exhibits low nanomolar cytotoxic activity against ERα+ breast cancer cells. It functions by activating the sodium channel TRPM4, which leads to an imbalance of intracellular calcium and sodium ions. This dysregulation of calcium homeostasis in ERα+ tumor cells triggers the anticipatory unfolded protein response and induces rapid immune cell-independent necrotic cell death. ErSO-TFPy is suitable for research on estrogen receptor alpha positive breast cancer.
  • ERα+ tumor cell inhibitor.
  • Exhibits low nanomolar cytotoxic activity against ERα+ breast cancer cells.
  • Activates the sodium channel TRPM4.
  • Causes an imbalance of intracellular calcium and sodium ions.
  • Dysregulates calcium homeostasis in ERα+ tumor cells.
  • Triggers the anticipatory unfolded protein response.
  • Induces rapid immune cell-independent necrotic cell death.
Catalog No. 50-003-72916