Learn More
Medchemexpress LLC Ertapenem sodium | 153773-82-1 | 98.9% | 497.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY13625
Encompass_Preferred
Ertapenem sodium (L-749345) is a broad-spectrum and long-acting β-lactam antibiotic that exhibits broad-spectrum anti-anaerobic activity.
  • Active against 99.1% of all anaerobes.
  • Suitable for research into severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • White to light yellow solid appearance.
  • Stable under recommended storage conditions.
Catalog No. 50-003-64905