Learn More
Medchemexpress LLC Ertapenem sodium | 153773-82-1 | MFCD09475566 | 97.4% | C22H24N3NaO7S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13625R10MG
Encompass_Preferred
Ertapenem sodium (Standard) is an analytical reference standard of the carbapenem antibiotic ertapenem, supplied as the sodium salt for research and analytical applications. It provides a well-characterized material for assay validation, method development, stability studies, and microbiological testing.
  • High purity analytical standard (97.4%) for reference and QC.
  • Broad-spectrum, long-acting β-lactam antibiotic with anti-anaerobic activity.
  • Molecular formula C22H24N3NaO7S and molecular weight 497.50.
  • Multiple pack sizes available to support small-scale and bulk testing.
  • Certificate of analysis and safety data sheet available for documentation.
Catalog No. 50-002-61165