Learn More
Medchemexpress LLC Ertapenem sodium (Standard) | 153773-82-1 | 97.4% | 497.50 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13625R
Encompass_Preferred
Ertapenem sodium (Standard) is the analytical standard of Ertapenem sodium. This product is intended for research and analytical applications. It is a broad-spectrum and long-acting β-lactam antibiotic with broad-spectrum anti-anaerobic activity against various anaerobes. It can be used for the research of severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-48819