Learn More
Medchemexpress LLC Erythorbic acid (standard) | 89-65-6 | MFCD00005378 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN7079R100MG
Encompass_Preferred
Erythorbic acid (standard), also known as D-Isoascorbic acid, is an analytical standard used for research and analytical applications. It is an orally active compound and a stereoisomer of L-ascorbic acid, primarily functioning as a food antioxidant.
  • Intended for research and analytical applications
  • Orally active compound
  • Improves the radiation resistance of Listeria monocytogenes
  • Reacts with free radicals and reduces oxidative damage
  • Possesses weak antiscorbic activity
  • Can prevent oxidative discoloration in meat products
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Functions as an antioxidant, reducing agent, and flavor enhancer
  • Used as an antimicrobial agent for food
Catalog No. 50-004-18948