Learn More
Medchemexpress LLC Erythrinin C | 63807-85-2 | 1 MG

Supplier: Medchemexpress LLC HYN38571MG
Erythrinin C is an isoflavone that can be isolated from *Erythrina suberosa* var. This compound presents as a white to off-white solid.
- Purity of 98%
- Molecular weight of 354.35
- Molecular formula C20H18O6
- CAS number 63807-85-2
- Isolated from plants, specifically Leguminosae, *Erythrina suberosa* Roxb.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.