Learn More
Selleck Chemical LLC Erythromycin S1635-1g
SDP
Supplier:  Selleck Chemical LLC S16351G
Encompass_Preferred
Erythromycin (E-Mycin) is a macrolide antibiotic that has an antimicrobial spectrum similar to or slightly wider than that of penicillin (IC50 1 5 g/ml)
Catalog No. 50-311-1137