Learn More
Cayman Chemical ErythromycIn-13C-d3 1mg
SDP
Supplier:  Cayman Chemical 253101 MG
Encompass_Preferred
An internal standard for the quantification of erythromycin by GC- or LC-MS
Catalog No. 50-293-7002