Learn More
Selleck Chemical LLC Erythromycin Ethylsuccinate S4060-50mg
SDP
Supplier:  Selleck Chemical LLC S406050MG
Encompass_Preferred
Erythromycin Ethylsuccinate an oral macrolide antibiotic produced by Streptomyces erythreus reversibly binds to the 50S ribosome of bacteria and inhibits protein synthesis
Catalog No. 50-310-8696