Learn More
Cayman Chemical ErythromycIn lactobIonate 5g
SDP
Supplier:  Cayman Chemical 171895 G
Encompass_Preferred
A macrolide antibiotic and water-soluble form of erythromycin; active against S. aureus and S. pneumoniae, as well as clinical isolates of group A streptococci (MICs = 0.6, 0.06, and 0.031 UG/ml, respectively); increases survival and reduces the number of lung CFUs in a mouse model of S. pneumoniae-induced pneumonia at 50 mg/kg; increases survival in mice infected with P. aeruginosa at 50 or 100 mg/kg
Catalog No. 50-296-6821