Learn More
Abcam Erythromycin, Macrolide antibiotic, 5G
Supplier:  Abcam AB1412055G
Encompass_Preferred
MW 733.9 Da. Macrolide antibiotic. Inhibits cytochrome p450 3A4 and P-glycoprotein. Binds the 50S ribosomal subunit and inhibits protein synthesis in Gram-positive and Gram-negative bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6801