Learn More
STEMCELL Technologies Erythropoietin (EPO) ELISA Kit, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 01630
Encompass
The glycoprotein EPO is the main physiological regulator of red blood cell formation. The EPO ELISA Kit is a rapid 3-step enzyme-linked immunosorbent assay (ELISA) designed for the quantitative measurement of natural and recombinant human EPO. The assay uses two monoclonal antibodies raised against human urinary EPO. These antibodies bind two non-overlapping epitopes on the EPO polypeptide and show high-affinity binding to both natural and recombinant EPO. The range of detection for the kit is 1.6 - 100 mU/mL. The EPO ELISA Kit has an assay time of approximately 3 hours.
Catalog No. NC1649435