Learn More
Medchemexpress LLC Erythrosin B sodium salt | 568-63-8 | MFCD00144257 | 85.0% | 879.86 g/mol | C20H6I4Na2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW20691125MG
Encompass_Preferred
Erythrosin B sodium salt is the sodium salt form of the xanthene-based dye erythrosin B. It is a water-soluble, red-colored iodinated dye widely used in laboratory settings as a staining reagent, biochemical assay dye, and photosensitizer for photochemical studies. Supplied as a solid for solution preparation, it is intended for research applications requiring consistent dye performance.
  • Water-soluble sodium salt, readily soluble in aqueous buffers.
  • Red fluorescent dye suitable for staining and visualization.
  • Used as a photosensitizer in photodynamic and photochemical studies.
  • Provided as a measured solid for reproducible solution preparation.
  • Commonly used in biochemical assays and histological staining.
Catalog No. 50-002-80785