Learn More
Selleck Chemical LLC ES9-17 E5911-25MG
SDP
Supplier:  Selleck Chemical LLC E591125MG
Encompass_Preferred
ES9-17 is a potent selective and reversible inhibitor of Clathrin-mediated endocytosis (CME) It bind to the N-terminal domain (nTD) of the clathrin heavy chain (CHC) inhibiting the uptake of FM4-64 and of several CME cargos in Arabidopsis
Catalog No. 50-000-41604