Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Esazomide impurity 11 | 69125-10-6 | C3H2ClN3O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY65092100MG

Encompass_Preferred

Esazomide impurity 11 is a chemical compound for laboratory research and the manufacture of substances. It serves as an impurity of Esazomide and is also used as a reference standard.

  • A chemical compound for laboratory use
  • Identified as an impurity of Esazomide
  • Molecular formula: C3H2ClN3O2
  • Molecular weight: 147.52
  • Appearance: Solid
  • Store container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Recommended storage at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-004-34776


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.