Learn More
Selleck Chemical LLC ESBA 105-A3098-1MG
SDP
Supplier:  Selleck Chemical LLC A30981MG
Encompass_Preferred
ESBA 105 is an antibody targeting tumor necrosis factor (TNF) It has an efficient intraocular penetration to anterior and posterior region of eye MW 145 14 KD
Catalog No. 50-313-9079