Learn More
ATCC Escherichia coli
Supplier:  ATCC 13706
Encompass
Escherichia coli strain C has applications in resistance testing and the detection of coliphages in water and it can be used as a bacteriophage host.
Catalog No. 50-238-2221