Learn More
ATCC Escherichia coli
Supplier:  ATCC 15669
Encompass
Escherichia coli strain HfrD was derived from Escherichia coli K-12 derivative Hfr Cavalli. This strain has lost the Hfr characteristic and is no longer a male-specific phage host.
Catalog No. 50-238-4383