Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers
Supplier:  ATCC 25922/DEL
Encompass
Escherichia coli strain Seattle 1946 is a whole-genome sequenced quality control strain that does not produce verotoxin. This organism is a CLSI control strain for antimicrobial susceptibility testing. It is used for media testing, as a negative control for LT toxin production, and as a quality control strain for Abbott, API, Autobac, BBL, bioMerieux VITEK, Biosynth, Difco, IDS, Micro-Media, MicroScan™, Roche Diagnostics, and Sensititre products. Used in susceptibility disc testing of neomycin, colistin [colimycin], kanamycin, cephalexin, gentamicins, cefamandole, cephalothin, tetracycline, cephaloglycin, cephaloridine [cephalomycin], nalidixic acid, and chloramphenicol.
Catalog No. 50-189-124FP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product