Learn More
ATCC Escherichia coli
Supplier:  ATCC 15597
Encompass
Escherichia coli strain C-3000 is a whole genome-sequenced strain that was derived from Escherichia coli K-12. The strain can be used as a bacteriophage host.
Catalog No. 50-238-1918