Learn More
ATCC Escherichia coli
Supplier:  ATCC 35218
Encompass
Escherichia coli strain 1532 was isolated from a canine in Tennessee. This whole-genome sequenced strain can be used in susceptibility disc testing or as a quality control strain.
Catalog No. 50-238-2848