Learn More
ATCC Escherichia coli
Supplier:  ATCC 25922
Encompass
Escherichia coli strain Seattle 1946 is a whole-genome sequenced quality control strain that does not produce verotoxin. This organism is a CLSI control strain for antimicrobial susceptibility testing. It is used for media testing, as a negative control for LT toxin production, and as a quality control strain for Abbott, API, Autobac, BBL, bioMerieux VITEK, Biosynth, Difco, IDS, Micro-Media, MicroScan, Roche Diagnostics, and Sensititre products. Used in susceptibility disc testing of neomycin, colistin [colimycin], kanamycin, cephalexin, gentamicins, cefamandole, cephalothin, tetracycline, cephaloglycin, cephaloridine [cephalomycin], nalidixic acid, and chloramphenicol.
Catalog No. 50-238-1849
 Restricted Item