Learn More
ATCC Escherichia coli; bacteriophage
Supplier:  ATCC 25404B1
Encompass
Escherichia coli bacteriophage P1 is a transducing phage that is propagated in Escherichia coli strain K-12 (ATCC 25404).
Catalog No. 50-238-4611
 Restricted Item