Learn More
ATCC Escherichia coli; S17-1
Supplier:  ATCC 47055
Encompass
Escherichia coli strain S17-1 is a recommended mobilization host for pARO180, pARO190, pARO181, and pARO191 (ATCC 77123-77126, respectively) because it contains chromosomally integrated tra genes.
Catalog No. 50-238-4242
 Restricted Item