Learn More
ATCC Escherichia coli; Strain 2006-3008
Supplier:  ATCC BAA2215
Encompass
Escherichia coli strain 2006-3008 is a whole-genome sequenced strain that was isolated in Idaho. It is toxigenic and was confirmed by PCR to carry the eae and stx1 genes. This strain is serotype O103:H11 and it has applications in food testing and enteric disease research.
Catalog No. 50-238-1912