Learn More
ATCC Escherichia coli; Strain 99-3311
Supplier:  ATCC BAA2192
Encompass
Escherichia coli strain 99-3311 is a whole-genome sequenced strain that was isolated from human stool in South Dakota. It is toxigenic and was confirmed by PCR to carry the stx1, stx2, and eae genes. It has applications for enteric disease research and food testing.
Catalog No. 50-238-3413