Learn More
Sigma Aldrich Fine Chemicals Biosciences Escherichia coli Strain B10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EC1130310G
Encompass
Escherichia coli is a non-pathogenic facultative anaerobic flora present in the intestine of human. It belongs to the genus Escherichia which consists of motile gram-negative bacilli.
Catalog No. 50-290-4271