Learn More
ATCC Escherichia coli; Strain M-200
Supplier:  ATCC 14169
Encompass
Escherichia coli strain NCIB 9270 [M-200] is a bacterium with applications in assays for vitamin B12.
Catalog No. 50-238-3541