Learn More
ATCC Escherichia coli; Strain O111
Supplier:  ATCC BAA2440
Encompass
Escherichia coli strain O111 is a whole-genome sequenced bacterium that can be used in enteric disease research, food testing, and infectious disease research. This strain is typed as serotype O111 and it is confirmed by PCR to contain the eae, stx1, and stx2 genes.
Catalog No. 50-238-4874